Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP - FAQs
Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP - FAQs
View additional product information for Goat anti-Mouse IgG (H+L) Secondary Antibody, HRP - FAQs (626520)
1 product FAQs found
What is the protein isoelectric point (PI) value of Goat anti-Mouse IgG (H+L) Secondary Antibody (Cat. No. 62-6520)?
We do not determine Kd or PI values for our antibodies.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.