Product Image
Thermo Scientific™
UltiMate™ 3000 Pump Filter Frit
Prevent contaminant particles from reaching your HPLC system by using these filter frits for your Thermo Scientific™ Dionex™ UltiMate™ 3000 Pumps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Material
6268.0111Dionex UltiMate 3000 RS Pump, Dionex UltiMate 3000 BM PumpTitanium
6268.0110Dionex UltiMate 3000 SD Pump, Dionex UltiMate 3000 RS PumpStainless Steel
6268.0117Ultimate 3000 BM PumpPEEK
Catalog number 6268.0111
Price (USD)
-
For Use With (Equipment):
Dionex UltiMate 3000 RS Pump, Dionex UltiMate 3000 BM Pump
Material:
Titanium
Prevent contaminant particles from reaching your HPLC system. Thermo Scientific™ Dionex™ UltiMate™ 3000 Pump Filter Frits remove contaminants that may be present in your solvent supply and prevent them from fouling your LC system. These frits are available in three different materials to match your chromatography needs.
Specifications
Biocompatible VersionYes
For Use With (Equipment)Dionex UltiMate 3000 RS Pump, Dionex UltiMate 3000 BM Pump
MaterialTitanium
TypeFilter Frit (10μm Porosity)
Unit SizeEach