Dynabeads™ DNA DIRECT™ Universal Kit
Invitrogen™

Dynabeads™ DNA DIRECT™ Universal Kit

The Dynabeads™ protocol consist of only a few steps of pipetting and magnetic separation, with all operations performed in aRead more
Have Questions?
Catalog NumberQuantity
63006300 Preps
Catalog number 63006
Price (USD)
735.65
Online exclusive
808.00
Save 72.35
Each
Quantity:
300 Preps
Price (USD)
735.65
Online exclusive
808.00
Save 72.35
Each
The Dynabeads™ protocol consist of only a few steps of pipetting and magnetic separation, with all operations performed in a single tube. Contaminating PCR inhibitors are eliminated without any centrifugation steps or use of phenol/chloroform. The kit is particularly useful for isolation of DNA from bacteria and cultured cells, as well as from clinical specimens and tissues from various species.

Benefits and Features:

• User-friendly 10 minute isolation of genomic DNA
• Only minute amount of starting material is required, even finger-prick samples can be used
• Protocols have been developed for fully automated use on liquid-handling robots, e.g. on the Biomek™ 2000, from Beckman Coulter, the Genesis RSP from Tecan, etc.

This Product Contains: The kit is supplied with Dynabeads™ and all neccesary buffers and solutions.

Applications: Isolation of PCR-ready genomic DNA directly from crude material
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
High-throughput CompatibilityNot High-throughput Compatible (Manual)
Product LineDNA DIRECT, DYNAL, Dynabeads
Product TypeUniversal Kit
Quantity300 Preps
Sample TypeFungi, Bacteria, Tissue, Blood, Cells, Buccal Samples
Shipping ConditionRoom Temperature
TargetGenomic DNA
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Frequently asked questions (FAQs)

I am seeing high PCR background when using the Dynabeads DNA DIRECT Kit. What could be the cause of this?

High PCR background could be caused by too high a concentration of template DNA, primers, Mg2+, or dNTPs. To fix this, reduce the amount of template DNA, primers, enzyme, and Mg2+ used, elute the DNA prior to PCR amplification, and/or try performing hot-start PCR. High background can also be caused by contamination. Ensure that the supernatant is completely removed at each washing step to avoid carryover.

I am getting inefficient elution of my DNA when using the Dynabeads DNA DIRECT Kit. Do you have any suggestions to improve this?

We recommend eluting the DNA in Resuspension Buffer, water, or low ionic strength buffer by incubation at 65 degrees C for > 5 mins. The complex must be fully resuspended before elution. The elution buffer can be pre-heated to 65 degrees C. After incubation, place tube in magnetic stand for 30 secs and transfer supernatant to a clean tube. To determine the elution efficiency, you can run some of the eluted DNA on an agarose gel. Alternatively, you can perform a PCR with the Dynabeads magnetic beads to determine if there is residual DNA remaining on the beads. Using more pre-heated elution buffer would be helpful to improve the elution efficiency.

I do not see any amplification when using your Dynabeads DNA DIRECT Kit for PCR. What should I do?

PCR may be inhibited by inhibitors, or an insufficient number of PCR cycles might have been used. For inhibitors, ensure that the washing buffer is brought to room temperature for use, and add the buffer more vigorously. You can also ensure that the supernatant is completely removed at each washing step, or introduce an additional washing step.

My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

I have a long double-stranded DNA fragment I would like to isolate. What product do you recommend?

For biotin-labled DNA that is less than 1 kb, we recommend you use Dynabeads M270 Streptavidin and MyOne C1 magnetic beads. We recommend our Dynabeads KilobaseBINDER Kit, which is designed to immobilize long (>1 kb) double-stranded DNA molecules. The KilobaseBINDER reagent consists of M-280 Streptavidin-coupled Dynabeads magnetic beads along with a patented immobilization activator in the binding solution to bind to long, biotinylated DNA molecules for isolation. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/immobilisation-of-long-biotinylated-dna-fragments.html) for more information in regards to long biotinylated DNA fragment isolation.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.