Immuno Clear Standard Modules
Immuno Clear Standard Modules
Immuno Clear Standard Modules
Immuno Clear Standard Modules
Thermo Scientific™

Immuno Clear Standard Modules

Perform applications that deliver well-to-well reproducibility with these immuno clear standard modules. Manufactured to fit standard equipment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSurface TreatmentVolume (Metric) WorkingWell ShapeWell Design
6302Immulon 2 HB330 μLRound, Flat Bottom (F-well)F12
434797MaxiSorp350 μLRound, Flat Bottom (F-well)F8
441653MaxiSorp250 μLRound, C BottomC8
444865PolySorp250 μLRound, C BottomC8
445101MaxiSorp250 μLRound, C BottomC8
467120MediSorp350 μLRound, Flat Bottom (F-well)F8
467140MultiSorp350 μLRound, Flat Bottom (F-well)F8
467466MaxiSorp350 μLRound, Flat Bottom (F-well)F16 (2x8 wells)
467679PolySorp350 μLRound, Flat Bottom (F-well)F16
468667MaxiSorp350 μLRound, Flat Bottom (F-well)F8
469078PolySorp350 μLRound, Flat Bottom (F-well)F8
469264Hydrophilic250 μLRoundU16
469914MaxiSorp350 μLRound, Flat Bottom (F-well)F16 (2x8 wells)
469922PolySorp350 μLRound, Flat Bottom (F-well)F16 (2x8 wells)
469949MaxiSorp350 μLRound, Flat Bottom (F-well)F8
469957PolySorp350 μLRound, Flat Bottom (F-well)F8
473709MaxiSorp250 μLRound, C BottomC12
473717PolySorp250 μLRound, C BottomC12
475078MaxiSorp250 μLRoundU8
475086PolySorp250 μLRoundU8
6309Immulon 2 HB330 μLRound, Flat Bottom (F-well)F12
6310Immulon 1 B330 μLRound, Flat Bottom (F-well)F12
6405Immulon 4 HBX330 μLRound, Flat Bottom (F-well)F12
95029100Enhanced Binding350 μLRound, Flat Bottom (F-well)F8
6301TS1
also known as 6301
Immulon 1 B330 μLRound, Flat Bottom (F-well)F12
Catalog number 6302
Price (USD)
231.32
Online exclusive
268.98
Save 37.66
Case of 320
Surface Treatment:
Immulon 2 HB
Volume (Metric) Working:
330 μL
Well Shape:
Round, Flat Bottom (F-well)
Well Design:
F12
Price (USD)
231.32
Online exclusive
268.98
Save 37.66
Case of 320

Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.

Features:

  • Several strip formats: Eight-well strip, 12-well strip, 16-well strip (two columns of eight wells)
  • Several well shapes: flat bottom, C bottom (radius edge), U shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules that remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm

Choice of Surface:

  • Thermo Scientific™ PolySorp™, Immulon™ 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific™ Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific™ MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins.

Ordering Information:

Modules are freestanding. Cat. No. 95029440 and 464394 are certified. Frames and modules are available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your customer service or sales representative for further details.

Specifications
Binding PropertySlightly Hydrophilic
Binding TypeAdsorption
BreakapartBreakable Strips
Certifications/ComplianceCertified for adsorption of IgG; CV of <5% between wells
CertifiedYes
Detection MethodColorimetric
FormatStrip Plate
LidWith Lid
No. per Case320
No. per Pack320
Packaging320 Pk., 320 Cases
Plate BlockingNone
Product TypeMicroplate Strip
Quantity320/Pk.
Target MoleculeProteins
Volume (Metric) Well380 μL
Volume (Metric) Working330 μL
Well DesignF12
ColorClear
MaterialPolystyrene
No. of Wells96
Surface TreatmentImmulon 2 HB
Well ShapeRound, Flat Bottom (F-well)
Unit SizeCase of 320

Frequently asked questions (FAQs)

What frames do you recommend for your Immuno Clear Standard Modules?

We recommend Frames and Accessories for Immuno Breakable Modules, BreakApart frame (Cat. No. 465404). This frame is designed to hold the Immuno Clear Standard Modules.

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.

Which Nunc immunodiagnostic passive surface should I choose: PolySorp, MediSorp, MaxiSorp, or MultiSorp?

Our passive immunodiagnostic surfaces are based on a series of treatments that modify the polystyrene surface to different final levels of hydrophilicity. Selection of one of these surfaces for ELISA development should be based on the characteristics of the biomolecule you seek to immobilize. PolySorp is fairly hydrophobic and should be used for biomolecules that have hydrophobic domains, e.g., lipids, lipoproteins, and large proteins. MediSorp is slightly hydrophilic and can be used for biomolecules with hydrophilic and hydrophobic properties, e.g., medium-to-large proteins such as albumin, or amphiphilic biomolecules such as LPS. MaxiSorp is quite hydrophilic, optimized for binding of IgG (and any other molecule with similar characteristics) and thus is a popular choice for sandwich ELISA and similar protocols. MultiSorp is very hydrophilic, and in turn will immobilize very hydrophilic biomolecules such as glycoproteins. Some optimization will always be necessary, and in many cases screening for binding optimization may involve a few surface types.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.

What is the difference between a certified and an uncertified immunodiagnostic plate, e.g., Cat. No. 439454 vs. 442404?

There is no difference in the manufacturing process. Certified plates are performance tested (using an antibody binding protocol) during quality control, whereas uncertified plates are not. This test is expensive for us to perform, so we offer the uncertified version at cost savings customers for whom antibody binding testing results are not critical.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.