Nalgene™ Polyolefin Pressure-Sensitive Labels
Nalgene™ Polyolefin Pressure-Sensitive Labels
Nalgene™ Polyolefin Pressure-Sensitive Labels
Nalgene™ Polyolefin Pressure-Sensitive Labels
Nalgene™ Polyolefin Pressure-Sensitive Labels
Thermo Scientific™

Nalgene™ Polyolefin Pressure-Sensitive Labels

Combine these waterproof, chemical resistant labels with photocopiers, laser printers or inkjet printers. Will not wrinkle, curl or melt.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)Width (Metric)QuantityUnit Size
6319-001022 mm22 mm4000 Cs.Case of 4000
6319-0010PK
also known as 6319-0010
22 mm22 mm800 Pk.Pack of 800
6319-0015PK
also known as 6319-0015
20 mm38 mm650 Pk.Pack of 650
6319-0020PK
also known as 6319-0020
25 mm67 mm300 Pk.Pack of 300
6319-0030PK13 mm44 mm100 Pk.Pack of 800
6319-0040PK51 mm102 mm100 Pk.Pack of 100
6319-001520 mm38 mm3250 Cs.Case of 3250
6319-002025 mm67 mm1500 Cs.Case of 1500
6319-003013 mm44 mm4000 Cs.Case of 4000
6319-004051 mm102 mm500 Cs.Case of 500
Catalog number 6319-0010
Price (USD)
647.10
Online exclusive
755.00
Save 107.90
Case of 4000
Height (Metric):
22 mm
Width (Metric):
22 mm
Quantity:
4000 Cs.
Unit Size:
Case of 4000
Price (USD)
647.10
Online exclusive
755.00
Save 107.90
Case of 4000

Combine Thermo Scientific™ Nalgene™ Polyolefin Pressure-Sensitive Labels with photocopiers, laser printers or inkjet printers. Waterproof and compatible with most lab chemicals, Nalgene polyolefin labels will not wrinkle, curl or melt. They accept a variety of inks including copier, marker, ballpoint, gels and technical markers.

Durable Polyolefin Labels

  • Rated to −29°C
  • Resistant to ultraviolet light, mildew, chemicals and dirt and won't discolor, crack or tear
  • Excellent for labeling microscopic slides— adhesive backing sticks well to any plastic, glass or metal surface
  • Chemically compatible with most lab reagents although solvents may dissolve adhesive
  • Autoclavable
  • Formatted for easy setup in word processing programs; instructions included in each pack

Note: For use with all writing devices except pencil. Test specific inks for suitability prior to use.

.

Specifications
DescriptionPolyolefin Label, 0.9 x 0.9
Height (English)0.9 in.
Height (Metric)22 mm
Labels per Sheet80
No. per Case4000/Cs.
No. per Pack800/Pk.
Product TypeLabel
Quantity4000 Cs.
Width (Metric)22 mm
MaterialPolyolefin
Width (English)0.9 in.
Unit SizeCase of 4000