Nalgene™ Polypropylene, Quick Action Spigot
Nalgene™ Polypropylene, Quick Action Spigot
Nalgene™ Polypropylene, Quick Action Spigot
Nalgene™ Polypropylene, Quick Action Spigot
Thermo Scientific™

Nalgene™ Polypropylene, Quick Action Spigot

For Use With: Nalgene carboys
Have Questions?
Catalog number 6422-0010
Price (USD)
500.38
Online exclusive
734.00
Save 233.62
Case of 12
Price (USD)
500.38
Online exclusive
734.00
Save 233.62
Case of 12

Stop drips and keep your Nalgene carboys in service for years with these Thermo Scientific™ Nalgene™ Polypropylene Quick-Action Replacement Spigots. Includes an HDPE tubing adapter which accepts tubing with 1/4 or 5/16 in. I.D for added versatility.

Features:

  • Spigot without tubing adapter accepts 5/8″ (1.6cm) I.D. tubing
  • Supplied with HDPE adapter for use with 1/4" or 5/16″ (0.6 or 0.8cm) tubing

Notes:

  • Disassemble spigot before autoclaving; tubing adapter is not autoclavable

Specifications
DescriptionQuick-Action Spigot PP fits 1-1/8 in. spigot boss for carboys 1-1/8 inch
Diameter (English) Inner Tubing5/8 in. (includes adapter for 1/4 or 5/16 in. tubing)
Diameter (Metric) Inner Tubing1.6 cm (includes adapter for 0.6 or 0.8 cm tubing)
For Use With (Equipment)Nalgene Carboys
No. per Pack1 Ea.
MaterialPolypropylene
Product TypeSpigot
Unit SizeCase of 12