Product Image
Invitrogen™

eBioscience™ Cell Proliferation Dye eFluor™ 450

Cell Proliferation Dye eFluor™ 450 is a violet fluorescent dye that can be used to monitor individual cell divisions. ThisRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
65-0842-904 x 500 μg
65-0842-85500 μg
Catalog number 65-0842-90
Price (USD)
415.65
Online exclusive
442.00
Save 26.35
Each
Quantity:
4 x 500 μg
Price (USD)
415.65
Online exclusive
442.00
Save 26.35
Each
Cell Proliferation Dye eFluor™ 450 is a violet fluorescent dye that can be used to monitor individual cell divisions. This fluorescent dye binds to any cellular proteins containing primary amines, and as cells divide, the dye is distributed equally between daughter cells that can be measured as successive halving of the fluorescence intensity of the dye. Up to 7 generations may be visualized. Cells labeled with Cell Proliferation Dye eFluor™ 450 may be fixed and permeabilized for analysis of intracellular targets using standard formaldehyde-containing fixatives and saponin-based permeabilization buffers, such as the Foxp3 Transcription Factor Staining Buffer Set (cat. 00-5523) or the IC Fixation Buffer (cat. 00-8222) and Permeabilization Buffer (10X) (cat. 00-8333).

Cell Proliferation Dye eFluor™ 450 has a peak excitation of 409 nm and can be detected using a 450/50 band pass filter (equivalent to eFluor™ 450 or Pacific Blue™), making it compatible with applications that utilize GFP.

Cell Proliferation Dye eFluor™ 450 is supplied as a lyophilized powder. Each vial may be reconstituted to a stock concentration of 10 mM with 165 μL of anhydrous DMSO; once reconstituted it should be used within 6 months and protected from light and stored at -20°C with dessicant. Avoid freeze-thawing.

Reported Application
Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Emission Wavelength405/450
FormLyophilized powder
Quantity4 x 500 μg
ColorViolet
For Use With (Application)Proliferation Assay
For Use With (Equipment)Flow Cytometer
Product LineeBioscience
Product TypeDye
Unit SizeEach