eBioscience™ Fixable Viability Dye eFluor™ 506, 100 Tests - FAQs
eBioscience™ Fixable Viability Dye eFluor™ 506, 100 Tests - FAQs
View additional product information for eBioscience™ Fixable Viability Dye eFluor™ 506 - FAQs (65-0866-14, 65-0866-18)
1 product FAQs found
Can Fixable Viability Dyes be used with the eBioscience BrdU Staining kits for Flow Cytometry?
Yes, a Fixable Viability Dye can be added to samples after BrdU labeling and before fixation with the BrdU Staining Buffer. See the protocol for additional information.