eBioscience™ Fixable Viability Dye eFluor™ 455UV, 100 tests - FAQs
eBioscience™ Fixable Viability Dye eFluor™ 455UV, 100 tests - FAQs
View additional product information for eBioscience™ Fixable Viability Dye eFluor™ 455UV - FAQs (65-0868-14, 65-0868-18)
2 product FAQs found
Can I use eBioscience Fixable Viability Dye eFluor 455UV (Cat. No. 65-0868-14 or 65-0868-18) in combination with Pacific Blue or eFluor 450?
Yes. The Fixable Viability Dye eFluor 455UV is excited by the UV laser and will not generate signal in the channels for dyes excited by the violet laser such as Pacific Blue and eFluor 450.
Can Fixable Viability Dyes be used with the eBioscience BrdU Staining kits for Flow Cytometry?
Yes, a Fixable Viability Dye can be added to samples after BrdU labeling and before fixation with the BrdU Staining Buffer. See the protocol for additional information.