eBioscience™ Fixable Viability Dye eFluor™ 455UV
Invitrogen™

eBioscience™ Fixable Viability Dye eFluor™ 455UV

Fixable Viability Dye eFluor™ 455UV is a viability dye that can be used to irreversibly label dead cells prior toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
65-0868-18500 tests
65-0868-14100 tests
Catalog number 65-0868-18
Price (USD)
473.65
Online exclusive
499.00
Save 25.35
Each
Quantity:
500 tests
Price (USD)
473.65
Online exclusive
499.00
Save 25.35
Each
Fixable Viability Dye eFluor™ 455UV is a viability dye that can be used to irreversibly label dead cells prior to cryopreservation, fixation and/or permeabilization procedures. Unlike 7-AAD and propidium iodide, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized, and stained for intracellular antigens without any loss of staining intensity of the dead cells. Thus, using Fixable Viability Dyes allows dead cells to be excluded from analysis when intracellular targets are being studied. Fixable Viability Dyes may be used to label cells from all species.

Fixable Viability Dye eFluor™ 455UV can be excited by a UV laser line (∼350 nm) and has a peak emission of 455 nm that can be detected using a 450/50 band pass filter (equivalent to DAPI). Please make sure that your instrument is capable of detecting this dye. For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye. If the percentage of dead cells is expected to be less than 5%, then it is recommended to take a small aliquot of cells and heat them at 65°C for 1 minute then immediately place on ice for 1 minute. After this treatment, the heat-killed cells can be combined 1:1 with live cells and then stained with the Fixable Viability Dye.

Fixable Viability Dye eFluor™ 455UV is supplied as a pre-diluted solution prepared in high-quality, anhydrous DMSO. It should be protected from light and moisture. Store at less than or equal to -70°C with dessicant. It may be freeze-thawed up to 20 times. Allow vial to equilibrate to room temperature before opening.

Reported Application
Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Emission Wavelength350/455
Quantity500 tests
ColorBlue
For Use With (Application)Viability Assay
For Use With (Equipment)Flow Cytometer
Product LineeBioscience
Product TypeDye
Unit SizeEach

Frequently asked questions (FAQs)

Can I use eBioscience Fixable Viability Dye eFluor 455UV (Cat. No. 65-0868-14 or 65-0868-18) in combination with Pacific Blue or eFluor 450?

Yes. The Fixable Viability Dye eFluor 455UV is excited by the UV laser and will not generate signal in the channels for dyes excited by the violet laser such as Pacific Blue and eFluor 450.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can Fixable Viability Dyes be used with the eBioscience BrdU Staining kits for Flow Cytometry?

Yes, a Fixable Viability Dye can be added to samples after BrdU labeling and before fixation with the BrdU Staining Buffer. See the protocol for additional information.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.