Product Image
Invitrogen™

eBioscience™ CyTRAK Orange™

CyTRAK Orange™ is an anthraquinone dye with high affinity for double-stranded DNA. It is a membrane-permeable dye that can labelRead more
Have Questions?
Catalog NumberQuantity
65-0881-9250 μL
Catalog number 65-0881-92
Price (USD)
239.65
Online exclusive
320.00
Save 80.35
Each
Quantity:
50 μL
Price (USD)
239.65
Online exclusive
320.00
Save 80.35
Each
CyTRAK Orange™ is an anthraquinone dye with high affinity for double-stranded DNA. It is a membrane-permeable dye that can label live or fixed/dead cells. In flow cytometry, it can be used to distinguish nucleated and non-nucleated cells. In fluorescent microscopy, it can be used to identify and discriminate the nucleus and cytoplasm without the need for a second dye due to its high intensity staining of the nucleus and low intensity staining of the cytoplasm.

CyTRAK Orange™ is optimally excited from 488 to 550 nm and has a peak emission of 610 nm. It can be excited with the blue laser (488 nm). For a filter configuration, CyTRAK Orange™ is best detected with a 610/20 band pass filter or equivalent. Please make sure that your instrument is capable of detecting this dye.

Reported Application
Microscopy, Flow Cytometric Analysis, Cell Labeling, Immunocytochemistry

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Quantity50 μL
ColorOrange
Emission488 to 550/610
For Use With (Application)Immunohistochemistry (IHC), Flow Cytometry
For Use With (Equipment)Fluorescence Microscope
Product LineeBioscience
Product TypeDye
Unit SizeEach