Versette™ Accessories, Pump module; Optional accessory, used for tip washing/reagent replenishing - FAQs
Versette™ Accessories, Pump module; Optional accessory, used for tip washing/reagent replenishing - FAQs
View additional product information for Versette™ Accessories - FAQs (650-04-PUMP)
5 product FAQs found
Which components for the Versette Automated Liquid Handler have serial numbers?
Serial numbers are listed on the three individual components which make up the complete instrument: base unit, six position stage, and 96/384 pipetting module. Additionally, the pipetting heads and the peristaltic pump have serial numbers for traceability.
What accessories are available for the Versette Automated Liquid Handler?
Here are the accessories available for the Versette Automated Liquid Handler:
- Three pipetting heads (volume range 0.5 µL - 300 µL)
- Peristaltic pump including non-contact reservoir fill
- Tip wash stations for 96/384-well formats
- Four serial dilution magazines for 96/384-well applications
What is the warranty on the Versette Automated Liquid Handler, including components, e.g., pipetting heads, pump, and tip wash station?
The warranty is 12 months from the date of shipment.
What liquid do you recommend using for washing pump tubing/fluid lines on the Versette Automated Liquid Handler?
Distilled/filtered laboratory grade water and/or a mild detergent solution, for example, filtered 1% Micro-90, followed by distilled and filtered laboratory-grade water is recommended to clean the pump tubing for the Versette Automated Liquid Handler.
Can reservoirs be automatically filled with the Versette Automated Liquid Handler?
Yes, a non-contact (N/C) reservoir fill module is included as an accessory with the purchase of a peristaltic pump (Cat. No. 650-04-PUMP).