Versette™ Accessories, 96 channel tip wash station, tall - FAQs
Versette™ Accessories, 96 channel tip wash station, tall - FAQs
View additional product information for Versette™ Accessories - FAQs (650-05-96TTW)
3 product FAQs found
What accessories are available for the Versette Automated Liquid Handler?
Here are the accessories available for the Versette Automated Liquid Handler:
- Three pipetting heads (volume range 0.5 µL - 300 µL)
- Peristaltic pump including non-contact reservoir fill
- Tip wash stations for 96/384-well formats
- Four serial dilution magazines for 96/384-well applications
What is the warranty on the Versette Automated Liquid Handler, including components, e.g., pipetting heads, pump, and tip wash station?
The warranty is 12 months from the date of shipment.
Can reservoirs be automatically filled with the Versette Automated Liquid Handler?
Yes, a non-contact (N/C) reservoir fill module is included as an accessory with the purchase of a peristaltic pump (Cat. No. 650-04-PUMP).