Versette™ Pipetting Heads, 96 channels, 5 to 300μL - FAQs
Versette™ Pipetting Heads, 96 channels, 5 to 300μL - FAQs
View additional product information for Versette™ Pipetting Heads - FAQs (650-06-96300)
5 product FAQs found
Which components for the Versette Automated Liquid Handler have serial numbers?
Serial numbers are listed on the three individual components which make up the complete instrument: base unit, six position stage, and 96/384 pipetting module. Additionally, the pipetting heads and the peristaltic pump have serial numbers for traceability.
What accessories are available for the Versette Automated Liquid Handler?
Here are the accessories available for the Versette Automated Liquid Handler:
- Three pipetting heads (volume range 0.5 µL - 300 µL)
- Peristaltic pump including non-contact reservoir fill
- Tip wash stations for 96/384-well formats
- Four serial dilution magazines for 96/384-well applications
What would happen if the wrong pipetting head is installed in the Versette Automated Liquid Handler?
The Versette Automated Liquid Handler incorporates RFID technology to identify which pipetting head is installed in the system. The protocol will prompt the user to check/change the head if the wrong head is installed for a specific protocol being run.
What is the warranty on the Versette Automated Liquid Handler, including components, e.g., pipetting heads, pump, and tip wash station?
The warranty is 12 months from the date of shipment.
Do I need any tools to change out the pipetting heads on the Versette Automated Liquid Handler?
No tools are required as the pipetting heads on the Versette Automated Liquid Handler can be changed through the ControlMate Software. This takes about 90 sec.