Versette™ Accessories, Serial dilute magazine 96/30μL (8/12) for use with short 30μL tips - FAQs
Versette™ Accessories, Serial dilute magazine 96/30μL (8/12) for use with short 30μL tips - FAQs
View additional product information for Versette™ Accessories - FAQs (650-08-9630SD)
1 product FAQs found
What accessories are available for the Versette Automated Liquid Handler?
Here are the accessories available for the Versette Automated Liquid Handler:
- Three pipetting heads (volume range 0.5 µL - 300 µL)
- Peristaltic pump including non-contact reservoir fill
- Tip wash stations for 96/384-well formats
- Four serial dilution magazines for 96/384-well applications