Versette™ ControlMate™ Software - FAQs
Versette™ ControlMate™ Software - FAQs
View additional product information for Versette™ ControlMate™ Software - FAQs (650-09-CMSW)
4 product FAQs found
Where can I find the Versette Automated Liquid Handler internal software version number?
The software version number for the Versette Automated Liquid Handler can be found in the following locations:
1) displayed on the onboard LCD screen System Configuration menu
2) displayed on the splash screen when opening the ControlMate Software
3) accessed in the HELP > About Menu
When using the Versette Automated Liquid Handler ControlMate Software, can I protect protocols from being changed/modified?
Yes, the Versette Automated Liquid Handler ControlMate Software has a specific feature to place edit and run locks on protocols via password protection.
How much do software updates typically cost for the Versette Automated Liquid Handler?
When software updates are available for the Thermo Scientific Versette Automated Liquid Handler, they can be accessed free of charge by visiting the Versette ControlMate Software product page (https://www.thermofisher.com/order/catalog/product/650-09-CMSW).
Is the software included with the purchase of the Versette Automated Liquid Handler? How many licenses do I get?
ControlMate Software for the Versette Automated Liquid Handler comes free with the purchase of the instrument; ControlMate Software is open licensed software for unlimited installation on multiple computers.