What frames do you recommend for your Immuno Clear Standard Modules?
We recommend Frames and Accessories for Immuno Breakable Modules, BreakApart frame (Cat. No. 465404). This frame is designed to hold the Immuno Clear Standard Modules.
Search
View additional product information for Immuno Clear Standard Modules - FAQs (6505TS)
1 product FAQs found