Dynabeads™ Streptavidin for Target Enrichment, 10 mL - FAQs
Dynabeads™ Streptavidin for Target Enrichment, 10 mL - FAQs
View additional product information for Dynabeads™ Streptavidin Magnetic Beads - FAQs (65604D, 65801D, 65605D, 65002, 65001, 65602, 65305, 65601, 11205D, 11206D, 65306, 65606D, 65607D, 60101, 60210)
1 product FAQs found
What range of DNA fragment sizes can be captured with Dynabeads Streptavidin for Target Enrichment (Cat. No. 65605D)?
We do not have hard cutoffs for the lengths of the fragments that can be captured with our Dynabeads products, including Cat. No. 65605D. In general terms, however, the beads should work well with any fragment up to 2kb. If you have mixed sizes in the sample, you can expect the smaller fragments being captured more efficiently than the longer ones.
Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.