Slide-A-Lyzer Float Buoys and Syringes are available to support a variety of dialysis cassettes for volumes of 0.1 to 20 mL.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Product Type
66490
5 mL
Cassette Syringe
66493
20 mL
Cassette Syringe
66494
1 mL
Cassette Syringe
3 Options
Catalog number 66490
Price (USD)
43.65
Online exclusive
50.50
Save 6.85
Each
Capacity (Metric):
5 mL
Product Type:
Cassette Syringe
Request bulk or custom format
Price (USD)
43.65
Online exclusive
50.50
Save 6.85
Each
Slide-A-Lyzer Dialysis Cassette Syringes are designed for efficient loading of samples into Slide-A-Lyzer cassettes and removal of sample after dialysis. Cassette Float Buoy accessories are for use with Slide-A-Lyzer Dialysis Cassettes used to remove low molecular weight contaminants and perform buffer exchange, desalting, and concentration.
Minimize sample loss into and out of Slide-A-Lyzer Dialysis Cassettes using Slide-A-Lyzer Dialysis Cassette Syringes for 1-mL to 20-mL sample volumes. Slide-A-Lyzer Cassette Float Buoys are designed for floating 0.5- to 12-mL cassettes in dialysate.
Features of Slide-A-Lyzer Cassette Accessories include: • Syringes are appropriately sized for different sample volumes • Buoys keep stir bar from tearing dialysis membrane • Matched to Slide-A-Lyzer Cassette for a secure fit and minimization of sample loss • Chemically inert
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity (Metric)5 mL
MembraneDialysis
Product LineSlide-A-Lyzer
Product TypeCassette Syringe
Quantity10 Syringes
Sufficient ForLoading samples into 3 mL Cassettes
FormatCassette, Accessory
Sample Volume (Metric)5 mL
Unit SizeEach
Contents & Storage
Store at room temperature.
Frequently asked questions (FAQs)
Does the Slide-A-Lyzer Dialysis Cassette leak?
The innovative, patented design does not permit wicking that can occur with homemade microdialysis devices.
What sizes are available for the Slide-A-Lyzer Dialysis Cassette? What MWCO membranes are available?
Four sizes to accommodate samples between 0.1 and 30 ml are currently available in five different MWCOs (2K, 3.5K, 7K, 10K and 20K).
The G2 Slide A lyzers on the other hand are available in five different sizes to accommodate samples between 0.25 and 70mL.
These units, especially those that accommodate smaller volumes, provide for greater sample recovery and shorter dialysis times than one can achieve with traditional tubing. We have a cassette selection guide to help you choose the appropriate cassette for your application. There are also product demonstrations for this and other High-Performance Dialysis Products.
What prevents the Slide-A-Lyzer Dialysis Cassette from leaking?
An internal gasket, which isn't visible after the cassette is assembled, reseals itself as the needle is withdrawn. To maintain sample volume and integrity, users should remember these three key rules:
1. Before use, fill the cassette with air to verify that the system is airtight.
2. Puncture each porthole only once.
3. Do not allow the membrane to come into contact with the sharp needle bevel.
What is the purpose of the buoy for the Slide-A-Lyzer Dialysis Cassette?
The buoy suspends the Slide-A-Lyzer Cassette above the stirring bar during dialysis and it provides a benchtop stand on which to set the Slide-A-Lyzer Cassette. These are not required with the G2 Slide-A-Lyzer Dialysis Cassettes.
What is the highest temperature and the pH range that is compatible with the Slide-A-Lyzer Cassette?
Slide-A-Lyzer Cassettes can be held at 56 degrees C for two hours. At temperatures and times above these, the plastic gets soft and can deform. Thermo Scientific recommends that users maintain a pH range of 5-9.