Rollout Drawers
Thermo Scientific™
Rollout Drawers
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire refrigerator shelving.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeCapacity (English)No. of Drawers
6700Complete SS Drawer Set51.1 cu. ft.12
6699Complete SS Drawer Set23.3 cu. ft.6
6702Additional SS Drawer78.8 cu. ft.1
6704Complete SS Drawer Set29.2 cu. ft.6
6725Additional SS Drawer29.2 cu. ft.1
7446Complete SS Drawer Set11.5 cu. ft.7
Catalog number 6700
Price (MXN)
-
Type:
Complete SS Drawer Set
Capacity (English):
51.1 cu. ft.
No. of Drawers:
12
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire shelving. This is an option for Thermo Scientific™ high-performance refrigerators.
  • Stainless-steel drawers
  • Adjustable with full extension
Ordering Information:
  • Factory installed; must be specified when ordering the refrigerator
Specifications
DrawersStainless Steel
For Use With (Equipment)51 cu. ft. (1447L) Refrigerators and Freezers
No. of Drawers12
TypeComplete SS Drawer Set
Capacity (English)51.1 cu. ft.
Capacity (Metric)1447 L
Unit SizeEach