Product Image
Gibco™
Resurge™ CD Pak
Resurge CD Pak are a diverse set of chemically defined supplement formulations to complement a variety of base media. The CD PAK contains one of each Resurge supplement: CD1, CD2, CD3, CD4, and CD5 at 100 grams each.
Have Questions?
Catalog NumberQuantity
6700301 Ea.
Catalog number 670030
Price (MXN)
-
Quantity:
1 Ea.
Resurge CD Pak are a diverse set of chemically defined supplement formulations to complement a variety of base media. The CD PAK contains one of each Resurge supplement: CD1, CD2, CD3, CD4, and CD5 at 100 grams each. The supplements have been developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. In order to increase versatility of the formulation, glucose and L-glutamine are not included in the final formulations. Therefore, these two components should be added by users and monitored as appropriate.
Specifications
Cell LineCHO
FormatBottles
Product LineResurge
Quantity1 Ea.
SourceAnimal origin-free
ClassificationAnimal Origin-free, Chemically-defined, Protein-free
FormPowder
Product TypeMedia Supplement
Unit SizeEach
Contents & Storage
•One of each Resurge supplement: CD1, CD2, CD3, CD4, and CD5 at 100 grams each.
•Storage conditions: 2° to 8°C.
•Store in cool, dry conditions in well-sealed containers.