Rollout Drawers
Thermo Scientific™
Rollout Drawers
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire refrigerator shelving.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeCapacity (English)No. of Drawers
6702Additional SS Drawer78.8 cu. ft.1
6699Complete SS Drawer Set23.3 cu. ft.6
6700Complete SS Drawer Set51.1 cu. ft.12
6704Complete SS Drawer Set29.2 cu. ft.6
6725Additional SS Drawer29.2 cu. ft.1
7446Complete SS Drawer Set11.5 cu. ft.7
Catalog number 6702
Price (USD)
-
Type:
Additional SS Drawer
Capacity (English):
78.8 cu. ft.
No. of Drawers:
1
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire shelving. This is an option for Thermo Scientific™ high-performance refrigerators.
  • Stainless-steel drawers
  • Adjustable with full extension
Ordering Information:
  • Factory installed; must be specified when ordering the refrigerator
Specifications
DrawersStainless Steel (Drawer)
For Use With (Equipment)23.3, 51.1 and 78.8 cu. ft. (659, 1447 and 2231L)
No. of Drawers1
TypeAdditional SS Drawer
Capacity (English)78.8 cu. ft.
Capacity (Metric)2231 L
Unit SizeEach