UltiMate™ AFC-3000 Collection Racks
Actual product may vary
UltiMate™ AFC-3000 Collection Racks
UltiMate™ AFC-3000 Collection Racks
UltiMate™ AFC-3000 Collection Racks
UltiMate™ AFC-3000 Collection Racks
UltiMate™ AFC-3000 Collection Racks
Thermo Scientific™
UltiMate™ AFC-3000 Collection Racks
Find the right rack for collecting your HPLC sample fractions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Capacity
6702.009090 Positions, 8 mL (13 mm O.D.)
6702.002121 Positions, 50 mL (33 mm O.D.)
6702.002424 Positions; 30 mL (26 mm O.D.)
6702.004040 Positions, 20 mL (21 mm O.D.)
6702.006060 Positions, 14 mL (15 mm O.D.)
Catalog number 6702.0090
Price (USD)
-
Sample Capacity:
90 Positions, 8 mL (13 mm O.D.)
Collect your samples for further analysis or storage in Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Collection Racks. The racks are available for a range of tube sizes from 8-50mL. The UltiMate AFC-3000 Automated Fraction Collector is a rugged and flexible fraction collector, ideal for preparative reversed-phase or normal phase HPLC as well as purification of proteins, peptides, and nucleotides.
Specifications
For Use With (Equipment)UltiMate AFC-3000 Fraction Collector
Sample Capacity90 Positions, 8 mL (13 mm O.D.)
TypeAFC-3000 Collection Rack
Unit SizeEach