Product Image
Thermo Scientific™

Buffer Tubing for UltiMate™ 3000 Autosamplers, SST

Select this stainless steel version of buffer tubing buffer tubing in volumes from 50 to 2000μL for your Thermo Scientific™ UltiMate™ 3000 Autosampler.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Capacity
6820.00561000 μL
6820.00212000 μL
6820.001950 μL
6820.0020500 μL
Catalog number 6820.0056
Price (USD)
-
Sample Capacity:
1000 μL

Choose Thermo Scientific™ Dionex™ Buffer Tubing for Thermo Scientific™ UltiMate™ 3000 Autosamplers, available in volumes from 50 to 2000μL. Select the stainless steel version of buffer tubing in the volume of your choice for your LC system.

Specifications
For Use With (Equipment)Ultimate 3000 Autosampler WPS-3000 TFC Analytical
MaterialStainless Steel
Sample Capacity1000 μL
TypeBuffer Tubing
Unit SizeEach