nano/Cap Modification Kits for UltiMate™ 3000 Autosamplers
Actual product may vary
Thermo Scientific™
nano/Cap Modification Kits for UltiMate™ 3000 Autosamplers
nano/Cap Modification Kits include 2.4 μL needle (fused silica, nanoViper), 25 μL syringe, 50 μL buffer loop, 1 μL sample loop, nanoViper, and 1 μL bridge (PEEKsil™) for WPS-3000TFC and for WPS-3000BTFC autosamplers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Biocompatible Version
6825.0030WPS-3000 TBFC AutosamplerYes
6824.0030WPS-3000 TFC AutosamplerNo
Catalog number 6825.0030
Price (USD)
-
For Use With (Equipment):
WPS-3000 TBFC Autosampler
Biocompatible Version:
Yes
Thermo Scientific™ Dionex™ nano/Cap Modification Kits include 2.4 μL needle (fused silica, nanoViper), 25 μL syringe, 50 μL buffer loop, 1 μL sample loop, nanoViper, and 1 μL bridge (PEEKsil™) for WPS-3000TFC and for WPS-3000BTFC autosamplers.
Specifications
DescriptionNano/Cap modification kit for WPS-3000 TBFC Analytical
Biocompatible VersionYes
For Use With (Equipment)WPS-3000 TBFC Autosampler
TypeNano/Cap Modification Kit
Unit SizeEach