CD3 Monoclonal Antibody (17A2), eFluor™ 506, eBioscience™ - FAQs
CD3 Monoclonal Antibody (17A2), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (17A2), eFluor™ 506, eBioscience™ - FAQs (69-0032-80, 69-0032-82)