CD3 Monoclonal Antibody (UCHT1), eFluor™ 506, eBioscience™ - FAQs
CD3 Monoclonal Antibody (UCHT1), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (UCHT1), eFluor™ 506, eBioscience™ - FAQs (69-0038-42, 69-0038-41)