CD4 Monoclonal Antibody (RPA-T4), eFluor™ 506, eBioscience™ - FAQs
CD4 Monoclonal Antibody (RPA-T4), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (RPA-T4), eFluor™ 506, eBioscience™ - FAQs (69-0049-41, 69-0049-42)