CD8a Monoclonal Antibody (53-6.7), eFluor™ 506, eBioscience™ - FAQs
CD8a Monoclonal Antibody (53-6.7), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (53-6.7), eFluor™ 506, eBioscience™ - FAQs (69-0081-82, 69-0081-80)