CD8a Monoclonal Antibody (RPA-T8), eFluor™ 506, eBioscience™ - FAQs
CD8a Monoclonal Antibody (RPA-T8), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (RPA-T8), eFluor™ 506, eBioscience™ - FAQs (69-0088-41, 69-0088-42)