CD11b Monoclonal Antibody (M1/70), eFluor™ 506, eBioscience™ - FAQs
CD11b Monoclonal Antibody (M1/70), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD11b Monoclonal Antibody (M1/70), eFluor™ 506, eBioscience™ - FAQs (69-0112-80, 69-0112-82)