CD11c Monoclonal Antibody (3.9), eFluor™ 506, eBioscience™ - FAQs
CD11c Monoclonal Antibody (3.9), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD11c Monoclonal Antibody (3.9), eFluor™ 506, eBioscience™ - FAQs (69-0116-42, 69-0116-41)