CD14 Monoclonal Antibody (61D3), eFluor™ 506, eBioscience™ - FAQs
CD14 Monoclonal Antibody (61D3), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD14 Monoclonal Antibody (61D3), eFluor™ 506, eBioscience™ - FAQs (69-0149-42, 69-0149-41)