CD19 Monoclonal Antibody (eBio1D3 (1D3)), eFluor™ 506, eBioscience™ - FAQs
CD19 Monoclonal Antibody (eBio1D3 (1D3)), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (eBio1D3 (1D3)), eFluor™ 506, eBioscience™ - FAQs (69-0193-82, 69-0193-80)