CD19 Monoclonal Antibody (HIB19), eFluor™ 506, eBioscience™ - FAQs
CD19 Monoclonal Antibody (HIB19), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (HIB19), eFluor™ 506, eBioscience™ - FAQs (69-0199-42, 69-0199-41)