CD45 Monoclonal Antibody (30-F11), eFluor™ 506, eBioscience™ - FAQs
CD45 Monoclonal Antibody (30-F11), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (30-F11), eFluor™ 506, eBioscience™ - FAQs (69-0451-80, 69-0451-82)