CD45R (B220) Monoclonal Antibody (RA3-6B2), eFluor™ 506, eBioscience™ - FAQs
CD45R (B220) Monoclonal Antibody (RA3-6B2), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD45R (B220) Monoclonal Antibody (RA3-6B2), eFluor™ 506, eBioscience™ - FAQs (69-0452-80, 69-0452-82)