CD45.2 Monoclonal Antibody (104), eFluor™ 506, eBioscience™ - FAQs
CD45.2 Monoclonal Antibody (104), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD45.2 Monoclonal Antibody (104), eFluor™ 506, eBioscience™ - FAQs (69-0454-80, 69-0454-82)