CD45RA Monoclonal Antibody (HI100), eFluor™ 506, eBioscience™ - FAQs
CD45RA Monoclonal Antibody (HI100), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD45RA Monoclonal Antibody (HI100), eFluor™ 506, eBioscience™ - FAQs (69-0458-42, 69-0458-41)