CD45 Monoclonal Antibody (HI30), eFluor™ 506, eBioscience™ - FAQs
CD45 Monoclonal Antibody (HI30), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (HI30), eFluor™ 506, eBioscience™ - FAQs (69-0459-41, 69-0459-42)