CD56 (NCAM) Monoclonal Antibody (TULY56), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD56 (NCAM) Monoclonal Antibody (TULY56), eFluor™ 506, eBioscience™ - FAQs (69-0566-42, 69-0566-41)