CD127 Monoclonal Antibody (eBioRDR5), eFluor™ 506, eBioscience™ - FAQs
CD127 Monoclonal Antibody (eBioRDR5), eFluor™ 506, eBioscience™ - FAQs
View additional product information for CD127 Monoclonal Antibody (eBioRDR5), eFluor™ 506, eBioscience™ - FAQs (69-1278-42)