Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 506, eBioscience™ - FAQs
Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 506, eBioscience™ - FAQs
View additional product information for Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 506, eBioscience™ - FAQs (69-4031-80, 69-4031-82)