Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 506, eBioscience™ - FAQs
Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 506, eBioscience™ - FAQs
View additional product information for Mouse IgG2a kappa Isotype Control (eBM2a), eFluor™ 506, eBioscience™ - FAQs (69-4724-80, 69-4724-82)