F4/80 Monoclonal Antibody (BM8), eFluor™ 506, eBioscience™ - FAQs
F4/80 Monoclonal Antibody (BM8), eFluor™ 506, eBioscience™ - FAQs
View additional product information for F4/80 Monoclonal Antibody (BM8), eFluor™ 506, eBioscience™ - FAQs (69-4801-82, 69-4801-80)