Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 506, eBioscience™ - FAQs
Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 506, eBioscience™ - FAQs
View additional product information for Armenian Hamster IgG Isotype Control (eBio299Arm), eFluor™ 506, eBioscience™ - FAQs (69-4888-80, 69-4888-82)