Ki-67 Monoclonal Antibody (SolA15), eFluor™ 506, eBioscience™ - FAQs
Ki-67 Monoclonal Antibody (SolA15), eFluor™ 506, eBioscience™ - FAQs
View additional product information for Ki-67 Monoclonal Antibody (SolA15), eFluor™ 506, eBioscience™ - FAQs (69-5698-82, 69-5698-80)