Ki-67 Monoclonal Antibody (20Raj1), eFluor™ 506, eBioscience™ - FAQs
Ki-67 Monoclonal Antibody (20Raj1), eFluor™ 506, eBioscience™ - FAQs
View additional product information for Ki-67 Monoclonal Antibody (20Raj1), eFluor™ 506, eBioscience™ - FAQs (69-5699-41, 69-5699-42)