FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 506, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 506, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (FJK-16s), eFluor™ 506, eBioscience™ - FAQs (69-5773-82, 69-5773-80)