KLRG1 Monoclonal Antibody (2F1), eFluor™ 506, eBioscience™ - FAQs
KLRG1 Monoclonal Antibody (2F1), eFluor™ 506, eBioscience™ - FAQs
View additional product information for KLRG1 Monoclonal Antibody (2F1), eFluor™ 506, eBioscience™ - FAQs (69-5893-82)